COMH580D8122 Combi-Blocks, Inc.

Methyl 4-chloro-8-fluoroquinoline-2-carboxylate

  • MDL number MFCD16036756
  • Cas Number : 219949-90-3
  • Molecular Weight : 239.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2321

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2321
Availability details for
Quantity
  • Availability
  • Add to Cart