COMH580D8124 Combi-Blocks, Inc.

4-Chloro-3-methylpyridine-2-carbaldehyde

  • Cas Number : 59886-86-1
  • Molecular Weight : 155.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4471
Availability details for
Quantity
  • Availability
  • Add to Cart