COMH580D8125 Combi-Blocks, Inc.

2,3-difluoro-4-nitrobenzonitrile

  • Cas Number : 1803870-92-9
  • Molecular Weight : 184.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4478

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4478
Availability details for
Quantity
  • Availability
  • Add to Cart