COMH580D8128 Combi-Blocks, Inc.

5-Bromo-1-N-ethyl-4-fluorobenzene-1,2-diamine

  • Cas Number : 1602586-39-9
  • Molecular Weight : 233.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4551
Availability details for
Quantity
  • Availability
  • Add to Cart