COMH580D812A Combi-Blocks, Inc.

{3-Methyl-1H-pyrazolo[3,4-b]pyridin-5-yl}boronic acid

  • MDL number MFCD22398789
  • Molecular Weight : 177.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4553
Availability details for
Quantity
  • Availability
  • Add to Cart