COMH580D8150 Combi-Blocks, Inc.

4-Bromo-1-{[2-(trimethylsilyl)ethoxy]methyl}pyrazole

  • MDL number MFCD26395073
  • Cas Number : 133560-58-4
  • Molecular Weight : 277.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole
Inchi Code 1S/C9H17BrN2OSi/c1-14(2,3)5-4-13-8-12-7-9(10)6-11-12/h6-7H,4-5,8H2,1-3H3
Inchi Key DIEAIDCZEZGMJP-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1488
Availability details for
Quantity
  • Availability
  • Add to Cart