COMH580D8153 Combi-Blocks, Inc.

5,6-Dimethoxy-1,3-dihydroindol-2-one

  • MDL number MFCD00221052
  • Cas Number : 6286-64-2
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0112

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0112
Availability details for
Quantity
  • Availability
  • Add to Cart