COMH580D8154 Combi-Blocks, Inc.

3-bromo-1-methylindole, tech grade

  • MDL number MFCD11102957
  • Cas Number : 81471-20-7
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0178

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0178
Availability details for
Quantity
  • Availability
  • Add to Cart