COMH580D8158 Combi-Blocks, Inc.

4-Chloro-5-nitrobenzene-1,2-dicarboxylic acid

  • Cas Number : 35010-28-7
  • Molecular Weight : 245.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1509
Availability details for
Quantity
  • Availability
  • Add to Cart