COMH580D815A Combi-Blocks, Inc.

6-chloro-N-cyclopropylpyridine-2-carboxamide

  • MDL number MFCD09862947
  • Cas Number : 149527-07-1
  • Molecular Weight : 196.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1948
Availability details for
Quantity
  • Availability
  • Add to Cart