COMH580D815B Combi-Blocks, Inc.

6-chloro-N-isopropylpyridine-2-carboxamide

  • Cas Number : 157328-17-1
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1949

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1949
Availability details for
Quantity
  • Availability
  • Add to Cart