COMH580D815F Combi-Blocks, Inc.

2-Bromo-1-cyclobutoxy-4-nitrobenzene

  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2486

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2486
Availability details for
Quantity
  • Availability
  • Add to Cart