COMH580D8160 Combi-Blocks, Inc.

4-Cyclobutoxybenzonitrile

  • MDL number MFCD16684468
  • Cas Number : 1249525-46-9
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2488
Availability details for
Quantity
  • Availability
  • Add to Cart