COMH580D8161 Combi-Blocks, Inc.

4-bromo-2-cyclobutoxy-1-nitrobenzene

  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2491

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2491
Availability details for
Quantity
  • Availability
  • Add to Cart