COMH580D8162 Combi-Blocks, Inc.

3-Bromo-4-methylbenzenecarbothioamide

  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2498

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2498
Availability details for
Quantity
  • Availability
  • Add to Cart