COMH580D8167 Combi-Blocks, Inc.

3-(benzyloxy)-2-bromo-5-chloropyridine

  • Molecular Weight : 298.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2515
Availability details for
Quantity
  • Availability
  • Add to Cart