COMH580D8169 Combi-Blocks, Inc.

5-Bromo-2-methyl-4-(4-nitrophenyl)-1,3-thiazole

  • MDL number MFCD28128058
  • Cas Number : 942920-44-7
  • Molecular Weight : 299.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2545
Availability details for
Quantity
  • Availability
  • Add to Cart