COMH580D8175 Combi-Blocks, Inc.

Esomeprazole magnesium trihydrate

  • MDL number MFCD07698573
  • Cas Number : 217087-09-7
  • Molecular Weight : 767.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?NA-0059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?NA-0059
Availability details for
Quantity
  • Availability
  • Add to Cart