COMH580D8178 Combi-Blocks, Inc.

N-[3-(Hydroxymethyl)phenyl]methanesulfonamide

  • MDL number MFCD12776133
  • Cas Number : 376347-09-0
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2326
Availability details for
Quantity
  • Availability
  • Add to Cart