COMH580D8179 Combi-Blocks, Inc.

2-(4-Fluorophenoxy)phenol

  • MDL number MFCD20441252
  • Cas Number : 91378-26-6
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-fluorophenoxy)phenol
Inchi Code 1S/C12H9FO2/c13-9-5-7-10(8-6-9)15-12-4-2-1-3-11(12)14/h1-8,14H
Inchi Key HEPMMEMCHCNTGU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2672
Availability details for
Quantity
  • Availability
  • Add to Cart