COMH580D817A Combi-Blocks, Inc.

1-[2-(Benzyloxy)phenyl]ethanamine hydrogen chloride

  • Molecular Weight : 263.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4309

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4309
Availability details for
Quantity
  • Availability
  • Add to Cart