COMH580D817C Combi-Blocks, Inc.

Cyclohexanol, 2-phenyl-

  • Cas Number : 1444-64-0
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-phenylcyclohexanol
Inchi Code 1S/C12H16O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-3,6-7,11-13H,4-5,8-9H2
Inchi Key AAIBYZBZXNWTPP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4791
Availability details for
Quantity
  • Availability
  • Add to Cart