COMH580D8180 Combi-Blocks, Inc.

2-Chloro-5-cyanobenzamide

  • Cas Number : 1261861-21-5
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4916

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4916
Availability details for
Quantity
  • Availability
  • Add to Cart