COMH580D8181 Combi-Blocks, Inc.

4-(Morpholin-4-yl)benzamide

  • MDL number MFCD22438486
  • Cas Number : 183557-77-9
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4918
Availability details for
Quantity
  • Availability
  • Add to Cart