COMH580D8182 Combi-Blocks, Inc.

tert-Butyl N-[(tert-butoxy)carbonyl]-N-(3-cyanophenyl)carbamate

  • Cas Number : 500024-52-2
  • Molecular Weight : 318.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (tert-butoxycarbonyl)(3-cyanophenyl)carbamate
Inchi Code 1S/C17H22N2O4/c1-16(2,3)22-14(20)19(15(21)23-17(4,5)6)13-9-7-8-12(10-13)11-18/h7-10H,1-6H3
Inchi Key UPFVGJRYWFWDNT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4919
Availability details for
Quantity
  • Availability
  • Add to Cart