COMH580D8184 Combi-Blocks, Inc.

3-(3-Bromophenoxy)oxane

  • Cas Number : 258523-55-6
  • Molecular Weight : 257.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-bromophenoxy)tetrahydro-2H-pyran
Inchi Code 1S/C11H13BrO2/c12-9-3-1-4-10(7-9)14-11-5-2-6-13-8-11/h1,3-4,7,11H,2,5-6,8H2
Inchi Key WCPPMSSSKRCJMJ-UHFFFAOYSA-N

Description

Purity 96%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4921

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4921
Availability details for
Quantity
  • Availability
  • Add to Cart