COMH580D81E5 Combi-Blocks, Inc.

3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethoxy)phenol

  • MDL number MFCD18756744
  • Cas Number : 1689502-03-1
  • Molecular Weight : 304.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5723
Availability details for
Quantity
  • Availability
  • Add to Cart