COMH580D81E6 Combi-Blocks, Inc.

2-[4-(tert-butylsulfanyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 909255-87-4
  • Molecular Weight : 292.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-(tert-butylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C16H25BO2S/c1-14(2,3)20-13-10-8-12(9-11-13)17-18-15(4,5)16(6,7)19-17/h8-11H,1-7H3
Inchi Key LOAPXNKARMOWAX-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5897

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5897
Availability details for
Quantity
  • Availability
  • Add to Cart