COMH580D81F0 Combi-Blocks, Inc.

Stannous chloride hydrate

  • MDL number MFCD00799154
  • Cas Number : 7772-99-8
  • Molecular Weight : 207.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0098
Availability details for
Quantity
  • Availability
  • Add to Cart