COMH580D81F1 Combi-Blocks, Inc.

Ethyl mesitylglyoxylate

  • MDL number MFCD00151821
  • Cas Number : 5524-57-2
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl mesityl(oxo)acetate
Inchi Code 1S/C13H16O3/c1-5-16-13(15)12(14)11-9(3)6-8(2)7-10(11)4/h6-7H,5H2,1-4H3
Inchi Key XRSPPFOBPRPYLD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0243

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0243
Availability details for
Quantity
  • Availability
  • Add to Cart