COMH580D81F2 Combi-Blocks, Inc.

Tin(iv) chloride n-hydrate

  • MDL number MFCD00216885
  • Cas Number : 7646-78-8
  • Molecular Weight : 278.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0270
Availability details for
Quantity
  • Availability
  • Add to Cart