COMH580D81F9 Combi-Blocks, Inc.

Hematoxylin hydrate

  • MDL number MFCD00150817
  • Cas Number : 517-28-2
  • Molecular Weight : 320.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1834
Availability details for
Quantity
  • Availability
  • Add to Cart