COMH580D8206 Combi-Blocks, Inc.

9-Fluorenone hydrazone

  • MDL number MFCD00016357
  • Cas Number : 13629-22-6
  • Molecular Weight : 194.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9H-fluoren-9-one hydrazone
Inchi Code 1S/C13H10N2/c14-15-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)13/h1-8H,14H2
Inchi Key YCNUILAKOMIBAL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4879
Availability details for
Quantity
  • Availability
  • Add to Cart