COMH580D820C Combi-Blocks, Inc.

5-(Dimethylaminomethyl)furfuryl alcohol hydrochloride

  • MDL number MFCD00040971
  • Cas Number : 81074-81-9
  • Molecular Weight : 191.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7358

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7358
Availability details for
Quantity
  • Availability
  • Add to Cart