COMH580D820E Combi-Blocks, Inc.

Zinc chloride hydrate

  • MDL number MFCD09880207
  • Cas Number : 7646-85-7
  • Molecular Weight : 154.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7660

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7660
Availability details for
Quantity
  • Availability
  • Add to Cart