COMH580D8218 Combi-Blocks, Inc.

Lobeline sulfate

  • MDL number MFCD00135592
  • Cas Number : 134-64-5
  • Molecular Weight : 773

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name sulfuric acid compound with 2-{(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methyl-2-piperidinyl}-1-phenylethanone (1:2)
Inchi Code 1S/2C22H27NO2.H2O4S/c2*1-23-19(15-21(24)17-9-4-2-5-10-17)13-8-14-20(23)16-22(25)18-11-6-3-7-12-18;1-5(2,3)4/h2*2-7,9-12,19-21,24H,8,13-16H2,1H3;(H2,1,2,3,4)/t2*19-,20+,21-;/m00./s1
Inchi Key GRZMOSSVIPFGFF-GNJLJDPWSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-8577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-8577
Availability details for
Quantity
  • Availability
  • Add to Cart