COMH580D8219 Combi-Blocks, Inc.

6-Iodo-2,3-dihydro-1,4-benzodioxine

  • MDL number MFCD00221460
  • Cas Number : 57744-67-9
  • Molecular Weight : 262.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9003
Availability details for
Quantity
  • Availability
  • Add to Cart