COMH580D821F Combi-Blocks, Inc.

Staurosporine

  • MDL number MFCD18252446
  • Cas Number : 220038-19-7
  • Molecular Weight : 466.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5S,6R,7R,9R)-6-methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one
Inchi Code 1S/C28H26N4O3/c1-28-26(34-3)17(29-2)12-20(35-28)31-18-10-6-4-8-14(18)22-23-16(13-30-27(23)33)21-15-9-5-7-11-19(15)32(28)25(21)24(22)31/h4-11,17,20,26,29H,12-13H2,1-3H3,(H,30,33)/t17-,20-,26-,28+/m1/s1
Inchi Key HKSZLNNOFSGOKW-FYTWVXJKSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1880
Availability details for
Quantity
  • Availability
  • Add to Cart