COMH580D8220 Combi-Blocks, Inc.

4-Chloro-n-(2-methoxybenzyl)-3-sulfamoylbenzamide

  • MDL number MFCD11726808
  • Cas Number : 1110762-81-6
  • Molecular Weight : 354.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3077
Availability details for
Quantity
  • Availability
  • Add to Cart