COMH580D8222 Combi-Blocks, Inc.

2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone

  • MDL number MFCD00799596
  • Cas Number : 33284-21-8
  • Molecular Weight : 175.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3282
Availability details for
Quantity
  • Availability
  • Add to Cart