COMH580D8225 Combi-Blocks, Inc.

Piperidine-4-carboxamidoxime

  • MDL number MFCD21117392
  • Cas Number : 1421266-74-1
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N'-hydroxy-4-piperidinecarboximidamide
Inchi Code 1S/C6H13N3O/c7-6(9-10)5-1-3-8-4-2-5/h5-6,8H,1-4,7H2
Inchi Key LKNINSSPPCSMQT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9041
Availability details for
Quantity
  • Availability
  • Add to Cart