COMH580D8229 Combi-Blocks, Inc.

Dibenzyl sulfoxide

  • MDL number MFCD00004782
  • Cas Number : 621-08-9
  • Molecular Weight : 230.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1058

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1058
Availability details for
Quantity
  • Availability
  • Add to Cart