COMH580D822B Combi-Blocks, Inc.

3-Phenyloxetane

  • MDL number MFCD17170019
  • Cas Number : 10317-13-2
  • Molecular Weight : 134.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2073
Availability details for
Quantity
  • Availability
  • Add to Cart