COMH580D822E Combi-Blocks, Inc.

Dimethyl acetamidomalonate

  • MDL number MFCD00066307
  • Cas Number : 60187-67-9
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4598
Availability details for
Quantity
  • Availability
  • Add to Cart