COMH580D8237 Combi-Blocks, Inc.

Ethyl 3-(bromomethyl)benzoate

  • MDL number MFCD13185929
  • Cas Number : 62290-17-9
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7611
Availability details for
Quantity
  • Availability
  • Add to Cart