COMH580D8239 Combi-Blocks, Inc.

1,8-Bis(methylamino)-3,6-dioxaoctane

  • MDL number MFCD00142574
  • Cas Number : 22366-98-9
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methyl-2-{2-[2-(methylamino)ethoxy]ethoxy}ethanamine
Inchi Code 1S/C8H20N2O2/c1-9-3-5-11-7-8-12-6-4-10-2/h9-10H,3-8H2,1-2H3
Inchi Key MRULVAADQTWBTF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8982

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8982
Availability details for
Quantity
  • Availability
  • Add to Cart