COMH580D823B Combi-Blocks, Inc.

Di-n-tfa-l-lysine methyl ester

  • MDL number MFCD00056343
  • Cas Number : 1478-74-6
  • Molecular Weight : 352.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (2S)-2,6-bis[(trifluoroacetyl)amino]hexanoate
Inchi Code 1S/C11H14F6N2O4/c1-23-7(20)6(19-9(22)11(15,16)17)4-2-3-5-18-8(21)10(12,13)14/h6H,2-5H2,1H3,(H,18,21)(H,19,22)/t6-/m0/s1
Inchi Key RENGOOCYAJRSCR-LURJTMIESA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9684

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9684
Availability details for
Quantity
  • Availability
  • Add to Cart