COMH580D823C Combi-Blocks, Inc.

1-Bromo-3-(trimethylsiloxy)benzene

  • MDL number MFCD00800273
  • Cas Number : 36971-28-5
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9798
Availability details for
Quantity
  • Availability
  • Add to Cart