COMH580D823F Combi-Blocks, Inc.

4'-Hydroxy-4-methoxychalcone

  • MDL number MFCD00020170
  • Cas Number : 6338-81-4
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6061
Availability details for
Quantity
  • Availability
  • Add to Cart