COMH580D8241 Combi-Blocks, Inc.

2-(4-Chlorophenoxy)pyridine-3-carbonyl chloride

  • MDL number MFCD00067837
  • Cas Number : 51362-50-6
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6466
Availability details for
Quantity
  • Availability
  • Add to Cart